trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals
trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number A15960.22
also known as A15960-22
Price (USD)/ Each
332.65
Online exclusive
369.00 
Save 36.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
332.65
Online exclusive
369.00 
Save 36.35 (10%)
Add to cart
trans-2-Octen-1-ol, 97%, Thermo Scientific Chemicals
Catalog numberA15960.22
Price (USD)/ Each
332.65
Online exclusive
369.00 
Save 36.35 (10%)
-
Add to cart
Chemical Identifiers
CAS18409-17-1
IUPAC Name(2E)-oct-2-en-1-ol
Molecular FormulaC8H16O
InChI KeyAYQPVPFZWIQERS-VOTSOKGWSA-N
SMILESCCCCC\C=C\CO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4440-1.4490 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only