Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: A15962.0B
1000 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-p-phenylenediamine, 96%, Thermo Scientific Chemicals

Catalog number: A15962.0B
1000 g, Each
Quantity
Catalog number: A15962.0B
also known as A15962-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-98-9
IUPAC Name
N1,N1-dimethylbenzene-1,4-diamine
Molecular Formula
C8H12N2
InChI Key
BZORFPDSXLZWJF-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(N)C=C1
Assay (GC)
≥95.0%
Appearance (Color)
Black or dark brown
Identification (FTIR)
Conforms
Form
Crystalline or fused solid
Melting Point (clear melt)
34-39?C

Description

N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

Solubility
Soluble in Water (11 g/L at20°C), diethyl ether, petroleum ether, alcohol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light and air sensitive. Store under inert gas. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text