Thermo Scientific Chemicals

n-Octyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15964.22
100 g, Each
Thermo Scientific Chemicals

n-Octyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A15964.22
100 g, Each
Quantity
Catalog number: A15964.22
also known as A15964-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1219-38-1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream
Melting Point (clear melt)
47.0-53.0?C
Assay (GC)
≥97.5%

Description

n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text