Thermo Scientific Chemicals

1,8-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A15966.18
50 g, Each
Thermo Scientific Chemicals

1,8-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: A15966.18
50 g, Each
Quantity
Catalog number: A15966.18
also known as A15966-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
479-27-6
IUPAC Name
naphthalene-1,8-diamine
Molecular Formula
C10H10N2
InChI Key
YFOOEYJGMMJJLS-UHFFFAOYSA-N
SMILES
NC1=CC=CC2=CC=CC(N)=C12
Appearance (Color)
Dark brown or purple
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Crystals, powder, crystalline lumps/powder or flakes
Melting Point
60.0-68.0?C

Description

1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Diaminonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text