Thermo Scientific Chemicals

2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A15974.06
5 g, Each
Thermo Scientific Chemicals

2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A15974.06
5 g, Each
Quantity
Catalog number: A15974.06
also known as A15974-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
769-25-5
IUPAC Name
2-ethenyl-1,3,5-trimethylbenzene
Molecular Formula
C11H14
InChI Key
PDELBHCVXBSVPJ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C=C)C(C)=C1
Solution Test
5% in diethyl ether: clear colorless
Assay (GC)
≥94.0%
Form
Liquid
Refractive Index
1.5265-1.5325 @ 20°C
Solution Test
5% in methanol: clear colorless

Description

2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

Solubility
Soluble in methanol.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text