Thermo Scientific Chemicals

Methyl cinnamate, predominantly trans, 99%, Thermo Scientific Chemicals

Catalog number: A15975.22
100 g, Each
Thermo Scientific Chemicals

Methyl cinnamate, predominantly trans, 99%, Thermo Scientific Chemicals

Catalog number: A15975.22
100 g, Each
Quantity
Catalog number: A15975.22
also known as A15975-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
103-26-4
IUPAC Name
methyl (2E)-3-phenylprop-2-enoate
Molecular Formula
C10H10O2
InChI Key
CCRCUPLGCSFEDV-BQYQJAHWSA-N
SMILES
COC(=O)\C=C\C1=CC=CC=C1
Appearance (Color)
Colorless to white
Form
Fused solid
Assay (GC)
≥98.5%
Melting Point (clear melt)
32.0-39.0?C

Description

Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text