3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals
3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A15981.06
also known as A15981-06
Price (USD)/ Each
36.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
36.60
Add to cart
3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals
Catalog numberA15981.06
Price (USD)/ Each
36.60
-
Add to cart
Chemical Identifiers
CAS620-24-6
IUPAC Name3-(hydroxymethyl)phenol
Molecular FormulaC7H8O2
InChI KeyOKVJCVWFVRATSG-UHFFFAOYSA-N
SMILESOCC1=CC=CC(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)64.0-75.0?C
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1'-acetoxy-1-methyl-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1′-acetoxy-1-methyl-benzol.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
A.Basu; S.S.Dixit; P.S.Phale. Metabolism of benzyl alcohol via catecholortho-pathway in methylnaphthalene-degrading Pseudomonas putida CSV86. Applied Microbiology and Biotechnology. 2003, 62, (5),579-585