Thermo Scientific Chemicals

3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A15981.14
25 g, Each
Thermo Scientific Chemicals

3-Hydroxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog number: A15981.14
25 g, Each
Quantity
Catalog number: A15981.14
also known as A15981-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
620-24-6
IUPAC Name
3-(hydroxymethyl)phenol
Molecular Formula
C7H8O2
InChI Key
OKVJCVWFVRATSG-UHFFFAOYSA-N
SMILES
OCC1=CC=CC(O)=C1
Appearance (Color)
White to cream to yellow to pale brown
Melting Point (clear melt)
64.0-75.0?C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%

Description

3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1'-acetoxy-1-methyl-benzol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1′-acetoxy-1-methyl-benzol.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text