3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1'-acetoxy-1-methyl-benzol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Hydroxybenzyl alcohol is used as an intermediate to produce other chemicals like 3-Oxy-1′-acetoxy-1-methyl-benzol.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
A.Basu; S.S.Dixit; P.S.Phale. Metabolism of benzyl alcohol via catecholortho-pathway in methylnaphthalene-degrading Pseudomonas putida CSV86. Applied Microbiology and Biotechnology. 2003, 62, (5),579-585