Thermo Scientific Chemicals

4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15985.36
500 g, Each
Thermo Scientific Chemicals

4,7-Dichloroquinoline, 98%, Thermo Scientific Chemicals

Catalog number: A15985.36
500 g, Each
Quantity
Catalog number: A15985.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
86-98-6
IUPAC Name
4,7-dichloroquinoline
Molecular Formula
C9H5Cl2N
InChI Key
HXEWMTXDBOQQKO-UHFFFAOYSA-N
SMILES
ClC1=CC=C2C(Cl)=CC=NC2=C1
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
82.0-88.0?C
Appearance (Color)
White to cream to yellow to pale brown

Description

4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,7-Dichloroquinoline is used as a drug chloroquine phosphate intermediate. It is also is used in the synthesis of hybrid aminoquinoline-triazine derivatives that show anti-microbial activity. In addition, it is used in the synthesis of novel oxazolidinones as anti-microbial agents showing efficacy against common bacterial strains.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text