Thermo Scientific Chemicals

trans,trans-2,4-Decadien-1-ol, 90%, remainder mainly trans, cis isomer, Thermo Scientific Chemicals

Catalog number: A15989.14
25 g, Each
Thermo Scientific Chemicals

trans,trans-2,4-Decadien-1-ol, 90%, remainder mainly trans, cis isomer, Thermo Scientific Chemicals

Catalog number: A15989.14
25 g, Each
Quantity
Catalog number: A15989.14
also known as A15989-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18409-21-7
IUPAC Name
(2E,4E)-deca-2,4-dien-1-ol
Molecular Formula
C10H18O
InChI Key
NUBWFSDCZULDCI-BLHCBFLLSA-N
SMILES
CCCCCC=CC=CCO
Appearance (Color)
Clear, colorless
Assay (GC)
>88.0%
Form
Liquid
Refractive Index
1.4855-1.4935 @ 20?C

Description

trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans,trans-2,4-Decadien-1-ol is used as food flavor and as an intermediate in organic chemical synthesis.

Solubility
Not miscible in water. Soluble in fat.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text