4-Fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number A15991.06
also known as A15991-06
Price (USD)/ Each
80.40
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
80.40
Add to cart
4-Fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberA15991.06
Price (USD)/ Each
80.40
-
Add to cart
Chemical Identifiers
CAS1765-93-1
IUPAC Name(4-fluorophenyl)boronic acid
Molecular FormulaC6H6BFO2
InChI KeyLBUNNMJLXWQQBY-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only