Thermo Scientific Chemicals

4,5-Dibromo-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

Catalog number: A15994.06
5 g, Each
Thermo Scientific Chemicals

4,5-Dibromo-3(2H)-pyridazinone, 98%, Thermo Scientific Chemicals

Catalog number: A15994.06
5 g, Each
Quantity
Catalog number: A15994.06
also known as A15994-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5788-58-9
IUPAC Name
4,5-dibromo-2,3-dihydropyridazin-3-one
Molecular Formula
C4H2Br2N2O
InChI Key
AGLQURQNVJVJNB-UHFFFAOYSA-N
SMILES
BrC1=C(Br)C(=O)NN=C1
Appearance (Color)
Orange to brown
Form
Powder
Assay (GC)
≥97.5%

Description

4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5-Dibromo-3(2H)-pyridazinone is used as an intermediate in the manufacture of pharmaceuticals. It is widely used in chemical research and in fine chemicals.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text