Thermo Scientific Chemicals

Theophylline monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A15997.0B
1000 g, Each
Thermo Scientific Chemicals

Theophylline monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A15997.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A15997.0B
also known as A15997-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5967-84-0
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥98.5% (U.S. sourced material)
Comment
Material Sourced in the U.S. and in other countries
Water Content (Karl Fischer Titration)
4.76-13.04% (0.5-1.5 waters, non-U.S. sourced material)
Form
Powder

Description

Theophylline monohydrate is a methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (8 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text