Theophylline monohydrate, 99%, Thermo Scientific Chemicals
Theophylline monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Theophylline monohydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A15997.30
also known as A15997-30
Price (USD)/ Each
82.70
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
82.70
Add to cart
Theophylline monohydrate, 99%, Thermo Scientific Chemicals
Catalog numberA15997.30
Price (USD)/ Each
82.70
-
Add to cart
Chemical Identifiers
CAS5967-84-0
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5% (non-U.S. sourced material)
Assay from Supplier's CofA≥98.5% (U.S. sourced material)
CommentMaterial Sourced in the U.S. and in other countries
Water Content (Karl Fischer Titration)4.76-13.04% (0.5-1.5 waters, non-U.S. sourced material)
FormPowder
View more
Theophylline monohydrate is a methylxanthine drug used in therapy for respiratory diseases such as COPD and asthma

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (8 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Naír Rodríguez-Hornedo.; David Lechuga-Ballesteros.; Hsiu-Jean Wu. Phase transition and heterogeneous/epitaxial nucleation of hydrated and anhydrous theophylline crystals. International Journal of Pharmaceutics. 1992, 85 (1-3), 149-162.
  2. Chamberlain O. Agbada.; Peter York. Dehydration of theophylline monohydrate powder — effects of particle size and sample weight. International Journal of Pharmaceutics. 1994, 106 (1), 33-40.