Thermo Scientific Chemicals

1,3-Dichloro-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: A16003.30
250 g, Each
Thermo Scientific Chemicals

1,3-Dichloro-2-propanol, 98+%, Thermo Scientific Chemicals

Catalog number: A16003.30
250 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16003.30
also known as A16003-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-23-1
IUPAC Name
1,3-dichloropropan-2-ol
Molecular Formula
C3H6Cl2O
InChI Key
DEWLEGDTCGBNGU-UHFFFAOYSA-N
SMILES
OC(CCl)CCl
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4810-1.4850 @ 20?C

Description

1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-2-propanol biotransformation to epichlorohydrin by the whole cells of recombinant Escherichia coli via resin-based in situ product removal has been investigated. It is used in the synthesis of glycerol, in the production of plastics and textiles and in the synthesis of pharmaceuticals. Also used as a solvent and as a cement for celluloid. It is also present in some foods, e.g. soy sauce, soup spices and instant soups, from hydrolization of proteins.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text