Thermo Scientific Chemicals

Lithium dodecylsulfate, 98%

Catalog number: A16005.14
25 g, Each
Thermo Scientific Chemicals

Lithium dodecylsulfate, 98%

Catalog number: A16005.14
25 g, Each
Quantity
Catalog number: A16005.14
also known as A16005-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2044-56-6
IUPAC Name
lithium(1+) dodecyl sulfate
Molecular Formula
C12H25LiO4S
InChI Key
YFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Appearance (Color)
White
Form
Powder
Assay (ex Sulphated Ash)
> 97.5%
Sulfated Ash
19.68 - 20.69%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text