Thermo Scientific Chemicals

cis-Aconitic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A16006.06
5 g, Each
Thermo Scientific Chemicals

cis-Aconitic anhydride, 98%, Thermo Scientific Chemicals

Catalog number: A16006.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16006.06
also known as A16006-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6318-55-4
IUPAC Name
2-(2,5-dioxo-2,5-dihydrofuran-3-yl)acetic acid
Molecular Formula
C6H4O5
InChI Key
GVJRTUUUJYMTNQ-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC(=O)OC1=O
Appearance (Color)
White to cream to pale brown
Form
Powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Infrared spectrum
Conforms
Melting Point (clear melt)
72-79?C

Description

cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic anhydride is used in the preparation of poly(amino acid)-containing copolymers with gradient pH-sensitive side groups initiated by the amino groups in methoxy poly(ethylene glycol)-block-poly(L-lysine). It is also involved in the preparation of pH-sensitive anti-tumor prodrug nanoparticles by conjugation with D-alfa-tocopheryl polyethylene glycol 1000 succinate (TPGS).

Notes
Moisture sensitive. Incompatible with strong acids, strong oxidizing agents and strong bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text