Thermo Scientific Chemicals

N-Boc-L-valine, 98+%, Thermo Scientific Chemicals

Catalog number: A16007.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-L-valine, 98+%, Thermo Scientific Chemicals

Catalog number: A16007.14
25 g, Each
Quantity
Catalog number: A16007.14
also known as A16007-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13734-41-3
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular Formula
C10H19NO4
InChI Key
SZXBQTSZISFIAO-ZETCQYMHSA-N
SMILES
CC(C)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
74.0-81.0?C
Optical Rotation
-7.4 ± 0.5? (C=1 in acetic acid)
Appearance (Color)
White

Description

N-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-valine is used in the multi-peptide synthesis and serves as the amino acid protection monomer.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text