Thermo Scientific Chemicals

Tetrazolium Violet, 98+%

Catalog number: A16008.03
1 g, Each
Thermo Scientific Chemicals

Tetrazolium Violet, 98+%

Catalog number: A16008.03
1 g, Each
Quantity
Catalog number: A16008.03
also known as A16008-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1719-71-7
IUPAC Name
2-(naphthalen-1-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Formula
C23H17ClN4
InChI Key
RONADMZTCCPLEF-UHFFFAOYSA-M
SMILES
[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Form
Powder
Appearance (Color)
Cream

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text