Chemical Identifiers
CAS1719-71-7
IUPAC Name2-(naphthalen-1-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular FormulaC23H17ClN4
InChI KeyRONADMZTCCPLEF-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=C2C=CC=CC2=CC=C1)C1=CC=CC=C1
View more