Thermo Scientific Chemicals

cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A16010.03
1 g, Each
Thermo Scientific Chemicals

cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals

Catalog number: A16010.03
1 g, Each
Quantity
Catalog number: A16010.03
also known as A16010-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
585-84-2
Appearance (Color)
White to cream to pale brown or pale orange
Assay (HPLC)
≥80.0% (major isomer)
Form
Powder

Description

cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

Solubility
Soluble in water, alcohol. Slightly soluble in ether.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text