Thermo Scientific Chemicals

Allyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A16012.14
25 g, Each
Thermo Scientific Chemicals

Allyltriphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog number: A16012.14
25 g, Each
Quantity
Catalog number: A16012.14
also known as A16012-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18480-23-4
IUPAC Name
triphenyl(prop-2-en-1-yl)phosphanium chloride
Molecular Formula
C21H20ClP
InChI Key
FKMJROWWQOJRJX-UHFFFAOYSA-M
SMILES
[Cl-].C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Melting Point (clear melt)
227.5-233.5?C
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text