Thermo Scientific Chemicals

N-Acetonylpyridinium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A16015.06
5 g, Each
Thermo Scientific Chemicals

N-Acetonylpyridinium chloride, 97%, Thermo Scientific Chemicals

Catalog number: A16015.06
5 g, Each
Quantity
Catalog number: A16015.06
also known as A16015-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42508-60-1
IUPAC Name
1-(2-oxopropyl)pyridin-1-ium chloride
Molecular Formula
C8H10ClNO
InChI Key
JHYYWKUENNZTMD-UHFFFAOYSA-M
SMILES
[Cl-].CC(=O)C[N+]1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
201.0-210.0?C
Appearance (Color)
Cream to pale brown
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetonylpyridinium chloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature. Protect from water and humidity.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text