Thermo Scientific Chemicals

4-Fluoro-2-methylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A16032.14
25 g, Each
Thermo Scientific Chemicals

4-Fluoro-2-methylphenol, 98%, Thermo Scientific Chemicals

Catalog number: A16032.14
25 g, Each
Quantity
Catalog number: A16032.14
also known as A16032-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
452-72-2
IUPAC Name
4-fluoro-2-methylphenol
Molecular Formula
C7H7FO
InChI Key
GKQDDKKGDIVDAG-UHFFFAOYSA-N
SMILES
CC1=CC(F)=CC=C1O
Appearance (Color)
Yellow
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
30.0-36.0?C
Assay (GC)
≥97.5%

Description

4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-2-methylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text