Thermo Scientific Chemicals

1,4-Phenylenediacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A16036.06
5 g, Each
Thermo Scientific Chemicals

1,4-Phenylenediacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A16036.06
5 g, Each
Quantity
Catalog number: A16036.06
also known as A16036-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7325-46-4
IUPAC Name
2-[4-(carboxylatomethyl)phenyl]acetate
Molecular Formula
C10H8O4
InChI Key
SLWIPPZWFZGHEU-UHFFFAOYSA-L
SMILES
[O-]C(=O)CC1=CC=C(CC([O-])=O)C=C1
Appearance (Color)
White to cream to pale yellow
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
249.0-258.0?C
Assay (Silylated GC)
≥96.0%

Description

1,4-Phenylenediacetic acid is an arenediacetic acid used in the preparation of flurorescent whitening agents. It has fluorescent properties that allow for use in fluorescent probes. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetic acid is an arenediacetic acid used in the preparation of flurorescent whitening agents. It has fluorescent properties that allow for use in fluorescent probes. It is also used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text