Thermo Scientific Chemicals

Indole-3-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A16041.03
1 g, Each
Thermo Scientific Chemicals

Indole-3-carbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A16041.03
1 g, Each
Quantity
Catalog number: A16041.03
also known as A16041-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5457-28-3
IUPAC Name
1H-indole-3-carbonitrile
Molecular Formula
C9H6N2
InChI Key
CHIFTAQVXHNVRW-UHFFFAOYSA-N
SMILES
N#CC1=CNC2=CC=CC=C12
Melting Point (clear melt)
175.5-181.5?C
Appearance (Color)
Cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-3-carbonitrile is used as a synthesis reagent for preparation of 4-substituted β-lactams, biologically active indoles, inhibitors of glycogen synthase kinase 3β (GSK-3), indole fragments as inosine monophosphate dehydrogenase (IMPDH) inhibitors and HIV-1 integrase inhibitors. It is also used as a reactant for intramolecular oxidative C-H coupling reactions with applications in medium-ring synthesis techniques and for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text