Thermo Scientific Chemicals

2-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A16042.06
5 g, Each
Thermo Scientific Chemicals

2-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: A16042.06
5 g, Each
Quantity
Catalog number: A16042.06
also known as A16042-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69360-26-5
IUPAC Name
2-cyanobenzene-1-sulfonyl chloride
Molecular Formula
C7H4ClNO2S
InChI Key
NQAYCMBZPAARNO-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=CC=C1C#N
Appearance (Color)
Pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Melting Point (clear melt)
63.5-69.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text