Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals
Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A16045.0E
also known as A16045-0E
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
Add to cart
Sodium percarbonate, 13-14% active oxygen, Thermo Scientific Chemicals
Catalog numberA16045.0E
Price (USD)/ Each
99.65
Online exclusive
111.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15630-89-4
IUPAC Nametetrasodium tris(peroxol) dicarbonate
Molecular FormulaC2H6Na4O12
InChI KeyMSLRPWGRFCKNIZ-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].OO.OO.OO.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or granular powder or granules
Available Oxygen12.5-14.5%
Assay (unspecified)(by Available Oxygen) : 81.8-94.8% assay
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium percarbonate is used as an oxidizing agent and environmentally friendly bleaching agent. It is used as a laboratory source of anhydrous hydrogen peroxide. It is utilized for the preparation of optically active 4-hydroxy-2-isoxazolines. It is widely used in detergent formulations and cleaning products such as carpet cleaner, textile bleach, denture cleaners, pulp and paper bleaching processes. It is also used as a hard filth removing agent in electroplating, waste water treatment agent and polymerization control agent. Further, it finds application in furniture polish and cleaners and personal care formulations. In addition to this, it is used in the preparation of trimethyl cyclopentenyl acetaldehyde and epoxydihydrolinalool.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong reducing agents, strong acids, organic materials and powdered metals.
RUO – Research Use Only

General References:

  1. Useful, inexpensive oxidant; reviews: Tetrahedron, 51, 6145 (1995); Synthesis, 1325 (1995); J. Chem. Soc., Perkin 1, 471 (2000). Examples: Conversion of nitriles to amides: Synth. Commun., 20, 1445 (1990). Cleavage of ɑ-diketones to acids: Synth. Commun., 23, 1183 (1993). Dakin or Baeyer-Villiger reaction (ultrasound-promoted): Tetrahedron Lett., 33, 865 (1992); or (in combination with TFA): Synthesis, 739 (1991). For an extensive review of the Baeyer-Villiger reaction, see: Org. React., 43, 251 (1993).
  2. Has been used in the presence of a catalytic amount of Pyridinium dichromate, L15132, or Potassium dichromate, A18722, and a phase-transfer catalyst, for the selective oxidation of secondary alcohols to ketones: Tetrahedron Lett., 35, 1989 (1994); Synth. Commun., 25, 2373 (1995).
  3. Can be used to generate the powerful oxidant peroxytrifluoroacetic acid from trifluoroacetic anhydride, avoiding the use of hazardous 90% hydrogen peroxide. The in situ generated peracid is useful in promoting the Baeyer-Villiger reaction of difficult substrates: Bull. Korean Chem. Soc., 17, 5 (1996).
  4. Nakano, M.; Wada, T.; Koga, N. Exothermic Behavior of Thermal Decomposition of Sodium Percarbonate: Kinetic Deconvolution of Successive Endothermic and Exothermic Processes. J. Phys. Chem. A 2015, 119 (38), 9761-9769.
  5. Hematian, S.; Hormozi, F. Drying kinetics of coated sodium percarbonate particles in a conical fluidized bed dryer. Powder Technol. 2015, 269, 30-37.