Thermo Scientific Chemicals

Triton X-100, Thermo Scientific Chemicals

Catalog number: A16046.AE
100 mL, Each
Thermo Scientific Chemicals

Triton X-100, Thermo Scientific Chemicals

Catalog number: A16046.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A16046.AE
also known as A16046-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-93-1
IUPAC Name
2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI Key
JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES
CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonym
triton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
Appearance (Color)
100 Max Pt-Co
Form
Liquid
Comment
Cloud Point: 63 - 70°C

Description

Detergent, equivalent to Nonidet P-40.Triton™ X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detergent, equivalent to Nonidet P-40.Triton|r X-100 is used as a non-ionic detergent, emulsifier and dispersing agent. It is useful for isolation of membrane proteins. It is also used in electrophoresis. It is an active ingredient in influenza vaccine (Fluzone). It finds applications in the dispersion of carbon materials for soft composite materials and restricting colony expansion in Aspergillus nidulans in microbiology. Further, it is an active ingredient in homemade vinyl record cleaning fluids together with distilled water and isopropyl alcohol.

Solubility
Miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text