Thermo Scientific Chemicals

Bis(2-ethylhexyl) sebacate, 95%, Thermo Scientific Chemicals

Catalog number: A16049.AK
250 mL, Each
Thermo Scientific Chemicals

Bis(2-ethylhexyl) sebacate, 95%, Thermo Scientific Chemicals

Catalog number: A16049.AK
250 mL, Each
Quantity
Catalog number: A16049.AK
also known as A16049-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-62-3
IUPAC Name
1,10-bis(2-ethylhexyl) decanedioate
Molecular Formula
C26H50O4
InChI Key
VJHINFRRDQUWOJ-UHFFFAOYNA-N
SMILES
CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC
Appearance (Color)
Clear colorless
Assay (GC)
≥94.0%
Refractive Index
1.4475-1.4535 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) sebacate is used as a plasticizer. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (0.1 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text