Thermo Scientific Chemicals

Diethylene glycol monomethyl ether, 98%, Thermo Scientific Chemicals

Catalog number: A16063.0F
2500 mL, Each
Thermo Scientific Chemicals

Diethylene glycol monomethyl ether, 98%, Thermo Scientific Chemicals

Catalog number: A16063.0F
2500 mL, Each
Quantity
Catalog number: A16063.0F
also known as A16063-0F
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-77-3
Refractive Index
1.4240 - 1.4280 @20?C
Assay (GC)
> 97.5%
Appearance (Color)
Clear, colourless
Form
liquid

Description

Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monomethyl ether is an industrial solvent and is also commonly used as a Fuel System Icing Inhibitor (FSII) in jet fuels.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text