Thermo Scientific Chemicals

Cyclohexane, 99+%, Thermo Scientific Chemicals

Catalog number: A16070.AU
1000 mL, Each
Thermo Scientific Chemicals

Cyclohexane, 99+%, Thermo Scientific Chemicals

Catalog number: A16070.AU
1000 mL, Each
Quantity
Catalog number: A16070.AU
also known as A16070-AU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-82-7
IUPAC Name
cyclohexane
Molecular Formula
C6H12
InChI Key
XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES
C1CCCCC1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms (UK-sourced material)
Refractive Index
1.4250-1.4275 @ 20?C (UK-sourced material)
Form
Liquid
Assay (GC)
≥99.0% (UK-sourced material)

Description

Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexane is used to prepare cyclohexanone-cyclohexanol mixture (i.e. KA oil) by catalytic oxidation, which finds application as raw material for adipic acid and caprolactam. It is used as an organic solvent, paint remover and involved in the calibration of differential scanning calorimetry (DSC) instruments.

Solubility
Miscible with ether, alcohol, acetone and olive oil. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text