Thermo Scientific Chemicals

alpha-Isopropylphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A16077.06
5 g, Each
Thermo Scientific Chemicals

alpha-Isopropylphenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: A16077.06
5 g, Each
Quantity
Catalog number: A16077.06
also known as A16077-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3508-94-9
IUPAC Name
3-methyl-2-phenylbutanoic acid
Molecular Formula
C11H14O2
InChI Key
HDLQGISFYDYWFJ-UHFFFAOYNA-N
SMILES
CC(C)C(C(O)=O)C1=CC=CC=C1
Appearance (Color)
White to pale yellow
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Melting Point (clear melt)
55.0-64.0?C
Form
Powder

Description

alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-Isopropylphenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text