Thermo Scientific Chemicals

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Catalog number: A16082.0E
2500 g, Each
Thermo Scientific Chemicals

1-Bromotetradecane, 98%, Thermo Scientific Chemicals

Catalog number: A16082.0E
2500 g, Each
Quantity
Catalog number: A16082.0E
also known as A16082-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-71-0
IUPAC Name
1-bromotetradecane
Molecular Formula
C14H29Br
InChI Key
KOFZTCSTGIWCQG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCBr
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4580-1.4620 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥97.5%

Description

1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4?-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromotetradecane was used in the synthesis of tetradecylferrocene, cationic and zwitterionic gemini surfactants, and 9-tetradecylcarbazole. Also used for synthesis of metallomesogenic polymers based on bis[η1(N)-3,4-dialkyloxybenzylidene-4′-dodecyloxyaniline]dichloropalladium(II) with octyl, decyl, dodecyl, tetradecyl and hexadecyl alkyl groups.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text