Thermo Scientific Chemicals

3-Methyl-2-buten-1-ol, 98+%, Thermo Scientific Chemicals

Catalog number: A16089.36
500 g, Each
Thermo Scientific Chemicals

3-Methyl-2-buten-1-ol, 98+%, Thermo Scientific Chemicals

Catalog number: A16089.36
500 g, Each
Quantity
Catalog number: A16089.36
also known as A16089-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
556-82-1
IUPAC Name
3-methylbut-2-en-1-ol
Molecular Formula
C5H10O
InChI Key
ASUAYTHWZCLXAN-UHFFFAOYSA-N
SMILES
CC(C)=CCO
Form
Liquid
Refractive Index
1.4410-1.4450 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-buten-1-ol is used as a reagent for protection of carboxylic acids as their 3-methyl-2-buten-1-yl (Prenyl) esters. It is also used in perfumery.

Solubility
Soluble in water (170 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text