Thermo Scientific Chemicals

PIPES, 98%, Thermo Scientific Chemicals

Catalog number: A16090.14
25 g, Each
Thermo Scientific Chemicals

PIPES, 98%, Thermo Scientific Chemicals

Catalog number: A16090.14
25 g, Each
Quantity
Catalog number: A16090.14
also known as A16090-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5625-37-6
IUPAC Name
sodium 2-[4-(2-sulfoethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular Formula
C8H17N2NaO6S2
InChI Key
OGGAIRCLBMGXCZ-UHFFFAOYSA-M
SMILES
[Na+].OS(=O)(=O)CCN1CCN(CCS([O-])(=O)=O)CC1
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

PIPES is commonly used as a buffering agent in biochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES is commonly used as a buffering agent in biochemistry.

Solubility
Soluble in 1M sodium hydroxide solution. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text