Thermo Scientific Chemicals

Acetamidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A16091.36
500 g, Each
Thermo Scientific Chemicals

Acetamidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: A16091.36
500 g, Each
Quantity
Catalog number: A16091.36
also known as A16091-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-42-5
IUPAC Name
hydrogen ethanimidamide chloride
Molecular Formula
C2H7ClN2
InChI Key
WCQOBLXWLRDEQA-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CC(N)=N
Form
Crystals or powder or crystalline powder or chunks
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Appearance (Color)
White to cream

Description

Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.

Solubility
Soluble in water.

Notes
It is hygroscopic. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text