Thermo Scientific Chemicals

BES, 99%, Thermo Scientific Chemicals

Catalog number: A16092.36
500 g, Each
Thermo Scientific Chemicals

BES, 99%, Thermo Scientific Chemicals

Catalog number: A16092.36
500 g, Each
Quantity
Catalog number: A16092.36
also known as A16092-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10191-18-1
IUPAC Name
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
Molecular Formula
C6H15NO5S
InChI Key
AJTVSSFTXWNIRG-UHFFFAOYSA-N
SMILES
OCCN(CCO)CCS(O)(=O)=O
Identification (FTIR)
Conforms (non-U.S. specification)
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

BES is used as a buffer to maintain the pH of the solutions in biological experiments. It is useful for diagnostic assay manufacturing industry. It induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes. It acts as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text