Thermo Scientific Chemicals

1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals

Catalog number: A16099.18
50 g, Each
Thermo Scientific Chemicals

1-(Trimethylsiloxy)cyclohexene, 98%, Thermo Scientific Chemicals

Catalog number: A16099.18
50 g, Each
Quantity
Catalog number: A16099.18
also known as A16099-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6651-36-1
IUPAC Name
(cyclohex-1-en-1-yloxy)trimethylsilane
Molecular Formula
C9H18OSi
InChI Key
SBEMOANGDSSPJY-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OC1=CCCCC1
Form
Liquid
Refractive Index
1.4445-1.4485 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless

Description

1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text