2-Ethoxyethanol, 99%, Thermo Scientific Chemicals
2-Ethoxyethanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethoxyethanol, 99%, Thermo Scientific Chemicals

Have Questions?
Quantity:
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16100.0F
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.30 
Save 13.65 (15%)
-
Add to cart
Quantity:
2500 mL
Request bulk or custom format
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.30 
Save 13.65 (15%)
Add to cart
2-Ethoxyethanol, 99%, Thermo Scientific Chemicals
Catalog numberA16100.0F
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
89.30 
Save 13.65 (15%)
-
Add to cart
Chemical Identifiers
CAS110-80-5
IUPAC Name2-ethoxyethan-1-ol
Molecular FormulaC4H10O2
InChI KeyZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILESCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5% (UK-sourced material)
Refractive Index1.4070-1.4100 @ 20?C (UK-sourced material)
FormLiquid
Assay from Supplier's CofA≥98.5% (US-sourced material)
View more
A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A widely used solvent for nitrocelluloses and dyes2-Ethoxyethanol is widely used as a varnish remover and degreasing solution. It is a solvent used for nitrocellulose. It is used as a component for dyes, inks, cleaning agents, resins, paints, destaining solution for lipoproteins on polyacrylamide gels.

Solubility
Miscible with water, acetone, ethyl acetate, benzene, carbon tetrachloride, diethyl ether, methanol, hydrocarbons and ethanol.

Notes
Incompatible with strong acids, strong bases, strong oxidizing agents, aluminum, alkalies and copper.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sedov, I. A.; Stolov, M. A.; Hart, E.; Grover, D.; Zettl, H.; Koshevarova, V.; William, A. E.; Abraham, M. H. Abraham model correlations for solute transfer into 2-ethoxyethanol from water and from the gas phase. J. Mol. Liq. 2015, 208, 63-70.
  2. Chiba, T.; Pu, Y. J.; Kido, J. Solution-Processed White Phosphorescent Tandem Organic Light-Emitting Devices. Adv. Mater. 2015, 27 (32), 4681-4687.