Undecanal, 97%, Thermo Scientific Chemicals
Undecanal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Undecanal, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A16101.14
also known as A16101-14
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
Add to cart
Undecanal, 97%, Thermo Scientific Chemicals
Catalog numberA16101.14
Price (USD)/ Each
33.65
Online exclusive
37.40 
Save 3.75 (10%)
-
Add to cart
Chemical Identifiers
CAS112-44-7
IUPAC Nameundecanal
Molecular FormulaC11H22O
InChI KeyKMPQYAYAQWNLME-UHFFFAOYSA-N
SMILESCCCCCCCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4315-1.4365 @ 20°C
View more
Undecanal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undecanal is used as a pharmaceutical intermediate.

Solubility
Soluble in dipropylene glycol, fixed oils, propylene glycol and water (14.27 mg/L at 25°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, reducing agents and bases. It is sensitive to air.
RUO – Research Use Only

General References:

  1. C.M Paleos Angelos Malliaris. Chemical evidence concerning the solubilization site of undecanal in micelles. Journal of Colloid and Interface Science. 1981, 82 (1), 244-247.
  2. George W. Mushrush, Robert N. Hazlett, Harold G. Eaton. Liquid-phase oxidation of undecanal by tert-butyl hydroperoxide in dodecane. Ind. Eng. Chem. Prod. Res. Dev. 1985, 24 (2), 290-293.