Thermo Scientific Chemicals

MES monohydrate, 98%

Catalog number: A16104.14
25 g, Each
Thermo Scientific Chemicals

MES monohydrate, 98%

Catalog number: A16104.14
25 g, Each
Quantity
Catalog number: A16104.14
also known as A16104-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
145224-94-8
IUPAC Name
4-(2-sulfonatoethyl)morpholin-4-ium
Molecular Formula
C6H13NO4S
InChI Key
SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CC[NH+]1CCOCC1
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5% (as monohydrate)
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES monohydrate is a biological buffer often referred to as Good’s buffer. It is useful for cell culture media and protein based buffer formulations to maintain a stable environment in solution. It is also used in the purification bioprocesses of antibodies, peptides, proteins, blood components and growth factors. It is a useful resolving agent for very small proteins on bis-tris gels. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text