Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16105.0E
2500 g, Each
Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: A16105.0E
2500 g, Each
Quantity
Catalog number: A16105.0E
also known as A16105-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-83-7
IUPAC Name
iron(3+) ammonium dodecahydrate disulfate
Molecular Formula
FeH28NO20S2
InChI Key
LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
White to purple
Form
Crystals or powder or crystalline powder
Assay (Iodometric Titration)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms

Description

Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) sulfate dodecahydrate is a reagent that is used as a mordant in dyeing and textile printing. It is also utilized in analytical chemistry as an iron standard. In biochemistry, ammonium ferric sulfate is used as a catalyst for the generation of free radicals. Increased xanthine oxidase and xanthine oxidoreductase activity in cultured rat cells has been demonstrated in the presence of ammonium ferric sulfate. It may be used as an iron source to restore enzymatic activity in apoenzymes, such as for soybean lipoxygenase.

Solubility
Soluble in water (1240 mg/ml at 25°C) and 1% (v/v) aqueous HCl (100 mg/ml). Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text