Thermo Scientific Chemicals

Cobalt(II,III) oxide, Thermo Scientific Chemicals

Catalog number: A16121.30
250 g, Each
Thermo Scientific Chemicals

Cobalt(II,III) oxide, Thermo Scientific Chemicals

Catalog number: A16121.30
250 g, Each
Quantity
Catalog number: A16121.30
also known as A16121-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-06-1
IUPAC Name
dicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular Formula
Co3O4
InChI Key
UBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
Total Metal Impurities
≤0.1% (U.S. specification)
Appearance (Color)
Fine grey to black
Complexometric Titration
Co content: 71-74% (Theory 73.42%, non-U.S. specification)
Form
Powder
Comment
Material specification is different for U.S. and non-U.S. purchases

Description

Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II,III) oxide is currently utilized in the process of artificial photosynthesis. It is also used for glass, optic and ceramic applications.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text