Thermo Scientific Chemicals

Betaine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16122.30
250 g, Each
Thermo Scientific Chemicals

Betaine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16122.30
250 g, Each
Quantity
Catalog number: A16122.30
also known as A16122-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-46-5
IUPAC Name
(carboxymethyl)trimethylazanium chloride
Molecular Formula
C5H12ClNO2
InChI Key
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
SMILES
[Cl-].C[N+](C)(C)CC(O)=O
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥98.5 to ≤101.5%
Form
Crystals or powder or crystalline powder or lumps

Description

Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Betaine hydrochloride is a naturally occurring compound that has been of interest for its role in osmoregulation. As a drug, it has been used as a source of hydrochloric acid in the treatment of hypochlorhydria. Betaine has also been used in the treatment of liver disorders, for hyperkalemia, for homocystinuria and for gastrointestinal disturbances.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text