Thermo Scientific Chemicals

L-Lysine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16127.22
100 g, Each
Thermo Scientific Chemicals

L-Lysine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: A16127.22
100 g, Each
Quantity
Catalog number: A16127.22
also known as A16127-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
657-26-1
IUPAC Name
(2S)-2,6-diaminohexanoic acid dihydrochloride
Molecular Formula
C6H16Cl2N2O2
InChI Key
JBBURJFZIMRPCZ-XRIGFGBMSA-N
SMILES
Cl.Cl.NCCCC[C@H](N)C(O)=O
Form
Powder or crystalline powder
Water Content (Karl Fischer Titration)
<3.50%
Optical Rotation
13-17? (c=2, in water)
Appearance (Color)
White to pale yellow
Assay (unspecified)
≥98.5 to ≤101.5% (dry wt. basis)

Description

Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hyperbranched polylysine using the N-hydroxysuccinimide ester of L-lysine dihydrochloride as AB 2 building block. The histidine and lysine standards were prepared from L-histidine monohydro- chloride and L-lysine dihydrochloride respectively. L-Homocitrulline-The copper complex was prepared from L-lysine dihydrochloride in 80 to 86 per cent yields.

Solubility
Soluble in water (100 mg/ml).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text