Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines, Thermo Scientific Chemicals
Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A16128.AP
also known as A16128-AP
Price (USD)/ Each
36.65
Online exclusive
41.00 
Save 4.35 (11%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
36.65
Online exclusive
41.00 
Save 4.35 (11%)
Add to cart
Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines, Thermo Scientific Chemicals
Catalog numberA16128.AP
Price (USD)/ Each
36.65
Online exclusive
41.00 
Save 4.35 (11%)
-
Add to cart
Chemical Identifiers
CAS112-24-3
IUPAC Name(2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine
Molecular FormulaC6H18N4
InChI KeyVILCJCGEZXAXTO-UHFFFAOYSA-N
SMILESNCCNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤150
Refractive Index1.4950-1.5010 @20°C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥55.0%
FormLiquid
Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only

General References:

  1. Michael F. Tweedle,; Lon J. Wilson.Variable spin iron(III) chelates with hexadentate ligands derived from triethylenetetramine and various salicylaldehydes. Synthesis, characterization, and solution state studies of a new 2T .dblarw. 6A spin equilibrium system. J. Am. Chem. Soc.. 1976, 98(16),4824-4834.
  2. Leandro Vinícius Alves Gurgel,; Laurent Frédéric Gil. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine. Water Research. 2009, 43 (18), 4479-4488.