Thermo Scientific Chemicals

D-Arginine, 98%

Catalog number: A16137.03
1 g, Each
Thermo Scientific Chemicals

D-Arginine, 98%

Catalog number: A16137.03
1 g, Each
Quantity
Catalog number: A16137.03
also known as A16137-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
157-06-2
IUPAC Name
(2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Formula
C6H14N4O2
InChI Key
ODKSFYDXXFIFQN-SCSAIBSYSA-N
SMILES
N[C@H](CCCN=C(N)N)C(O)=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Optical Rotation
-26.5 ± 1.0? (C=1.6 in 5N HCl)

Description

1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand's disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Deamino-8-D-arginine vasopressin makes a new pharmacological approach to the management of haemophilia and von Willebrand’s disease. A novel nonenzymatic pathway for the generation of nitric oxide is by the reaction of hydrogen peroxide and d- or l-arginine.

Solubility
Insoluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text