(+/-)-3-Chloro-1,2-propanediol, 98%, Thermo Scientific Chemicals
(+/-)-3-Chloro-1,2-propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(+/-)-3-Chloro-1,2-propanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
500 g
Catalog number A16143.0E
also known as A16143-0E
Price (USD)/ Each
142.00
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
142.00
Add to cart
(+/-)-3-Chloro-1,2-propanediol, 98%, Thermo Scientific Chemicals
Catalog numberA16143.0E
Price (USD)/ Each
142.00
-
Add to cart
Chemical Identifiers
CAS96-24-2
IUPAC Name3-chloropropane-1,2-diol
Molecular FormulaC3H7ClO2
InChI KeySSZWWUDQMAHNAQ-UHFFFAOYNA-N
SMILESOCC(O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4780-1.4820 @ 20*C
FormLiquid or viscous liquid
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only